Merck
DEWAX Transcription Factor Is Involved in Resistance to
Frontiers in plant science (2017-07-27)
Seulgi Ju, Young Sam Go, Hyo Ju Choi, Jeong Mee Park, Mi Chung Suh
ABSTRACT
The cuticle of land plants is the first physical barrier to protect their aerial parts from biotic and abiotic stresses. DEWAX, an AP2/ERF-type transcription factor, negatively regulates cuticular wax biosynthesis. In this study, we investigated the resistance to
Sigma-Aldrich
Sodium 3,5-dichloro-2-hydroxybenzenesulfonate, used for peroxide measurement