Merck
  • Dual Stimuli-Responsive Nanoparticles for Controlled Release of Anticancer and Anti-inflammatory Drugs Combination.
Dual Stimuli-Responsive Nanoparticles for Controlled Release of Anticancer and Anti-inflammatory Drugs Combination.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-05-11)
Liandong Feng, Yuqi Wang, Zhiliang Luo, Zheng Huang, Yan Zhang, Kai Guo, Deju Ye
ABSTRACT
Dual stimuli-responsive nanoparticles capable of fine-tuning drug release to augment therapeutic efficacy have become a promising tool for anticancer drug delivery. However, the rational design of these "smart" nanoparticles for a selective delivery and controlled release of multidrug combinations in cancer cells to achieve synergistic effects remain challenging. Here we report the pH/redox dual responsive nanoparticle FA-DOX-Ind-NP (FA=folic acid, DOX=doxorubicin, Ind=indomethacin, NP=nanoparticle) based on the novel tumor targeting and biodegradable poly(β-amino ester) polymer, and demonstrate its high ability to enter into cancer cells and release a combination of the anticancer drug doxorubicin and the non-steroidal anti-inflammatory drug indomethacin to achieve synergistic chemo-anti-inflammatory effects and overcome multidrug resistance. This study highlights the great potential of tumor targeting and dual stimuli-responsive nanoparticles for an efficient delivery of multidrug combination to improve the cancer therapeutic efficacy.
Sigma-Aldrich
Tetradecylamine, 95%