- CD74 Deficiency Mitigates Systemic Lupus Erythematosus-like Autoimmunity and Pathological Findings in Mice.
CD74 Deficiency Mitigates Systemic Lupus Erythematosus-like Autoimmunity and Pathological Findings in Mice.
Journal of immunology (Baltimore, Md. : 1950) (2017-02-22)
Yi Zhou, Huimei Chen, Li Liu, Xueqing Yu, Galina K Sukhova, Min Yang, Lijun Zhang, Vasileios C Kyttaris, George C Tsokos, Isaac E Stillman, Takaharu Ichimura, Joseph V Bonventre, Peter Libby, Guo-Ping Shi
PMID28219888
ABSTRACT
CD74 mediates MHC class-II antigenic peptide loading and presentation and plays an important role in the pathogenesis of autoimmune diseases, including systemic lupus erythematosus. C57BL/6
Sigma-Aldrich
Deoxyribonucleic acid−cellulose double-stranded from calf thymus DNA, lyophilized powder