Merck
  • Resveratrol Reactivates Latent HIV through Increasing Histone Acetylation and Activating Heat Shock Factor 1.
Resveratrol Reactivates Latent HIV through Increasing Histone Acetylation and Activating Heat Shock Factor 1.
Journal of agricultural and food chemistry (2017-05-05)
Xiaoyun Zeng, Xiaoyan Pan, Xinfeng Xu, Jian Lin, Fuchang Que, Yuanxin Tian, Lin Li, Shuwen Liu
ABSTRACT
The persistence of latent HIV reservoirs presents a significant challenge to viral eradication. Effective latency reversing agents (LRAs) based on "shock and kill" strategy are urgently needed. The natural phytoalexin resveratrol has been demonstrated to enhance HIV gene expression, although its mechanism remains unclear. In this study, we demonstrated that resveratrol was able to reactivate latent HIV without global T cell activation in vitro. Mode of action studies showed resveratrol-mediated reactivation from latency did not involve the activation of silent mating type information regulation 2 homologue 1 (SIRT1), which belonged to class-3 histone deacetylase (HDAC). However, latent HIV was reactivated by resveratrol mediated through increasing histone acetylation and activation of heat shock factor 1 (HSF1). Additionally, synergistic activation of the latent HIV reservoirs was observed under cotreatment with resveratrol and conventional LRAs. Collectively, this research reveals that resveratrol is a natural LRA and shows promise for HIV therapy.
Sigma-Aldrich
Triacetyl resveratrol, ≥98% (HPLC)
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
SAHA, ≥98% (HPLC)