Merck
  • EBI2 Expression and Function: Robust in Memory Lymphocytes and Increased by Natalizumab in Multiple Sclerosis.
EBI2 Expression and Function: Robust in Memory Lymphocytes and Increased by Natalizumab in Multiple Sclerosis.
Cell reports (2017-01-05)
Aurélie S Clottu, Amandine Mathias, Andreas W Sailer, Myriam Schluep, Jörg D Seebach, Renaud Du Pasquier, Caroline Pot
ABSTRACT
The interaction between oxysterols and the G protein-coupled receptor Epstein-Barr virus-induced gene 2 (EBI2) fine-tunes immune cell migration, a mechanism efficiently targeted by several disease-modifying treatments developed to treat multiple sclerosis (MS), such as natalizumab. We previously showed that memory CD4
Sigma-Aldrich
NIBR189, ≥98% (HPLC)