Merck
  • Efficient transfection of Xenobiotic Responsive Element-biosensor plasmid using diether lipid and phosphatidylcholine liposomes in differentiated HepaRG cells.
Efficient transfection of Xenobiotic Responsive Element-biosensor plasmid using diether lipid and phosphatidylcholine liposomes in differentiated HepaRG cells.
International journal of pharmaceutics (2017-04-04)
Maxime Demazeau, Nicolas Quesnot, Nicolas Ripoche, Claudine Rauch, Jelena Jeftić, Fabrice Morel, Fabienne Gauffre, Thierry Benvegnu, Pascal Loyer
ABSTRACT
In this study, we evaluated cationic liposomes prepared from diether-NH
Product Description
Supelco
Endosulfan, PESTANAL®, analytical standard
Sigma-Aldrich
HepaRG Induction Medium Supplement with Antibiotics
Sigma-Aldrich
HepaRG Thawing and Plating Medium Supplement with Antibiotics
Sigma-Aldrich
HepaRG Maintenance and Metabolism Medium Supplement with Antibiotics
Sigma-Aldrich
HepaRG Serum Free Induction Medium Supplement
Sigma-Aldrich
HepaRG Thawing and Plating Medium Supplement
Sigma-Aldrich
HepaRG Maintenance and Metabolism Medium Supplement
Sigma-Aldrich
HepaRG Induction Medium Supplement
Sigma-Aldrich
HepaRG Serum Free Induction Medium Supplement with Antibiotics