Merck
  • Mitochondrial phosphatase PGAM5 regulates Keap1-mediated Bcl-xL degradation and controls cardiomyocyte apoptosis driven by myocardial ischemia/reperfusion injury.
Mitochondrial phosphatase PGAM5 regulates Keap1-mediated Bcl-xL degradation and controls cardiomyocyte apoptosis driven by myocardial ischemia/reperfusion injury.
In vitro cellular & developmental biology. Animal (2016-11-07)
Chen Yang, Xiaojuan Liu, Fangfang Yang, Weixun Zhang, Zihao Chen, Daliang Yan, Qingsheng You, Xiang Wu
ABSTRACT
Phosphoglycerate mutase 5 (PGAM5) is a mitochondrial membrane protein that plays crucial roles in necroptosis and apoptosis. Though PGAM5 is known to be required for inducing intrinsic apoptosis through interacting with BCL2 associated X protein (Bax) and dynamin-related protein 1 (Drp1), the expression and role of PGAM5 in cardiomyocyte apoptosis driven by myocardial ischemia/reperfusion injury(MIRI) has not been studied. The present study shows that PGAM5 expression decreased after MIRI in vivo, positively correlated with Bcl-xL expression, negatively correlated with Kelch-ECH associating protein 1 (Keap1) expression. Furthermore, PGAM5 expression also decreased in cardiomyocytes after hypoxia/reoxygenation (H/R) treatment in vitro. PGAM5 silence promoted cardiomyocyte apoptosis and inhibited Bcl-xL expression, but with no effect on Keap1 expression. Accordingly, Keap1 overexpression further inhibited Bcl-xL and PGAM5 expression. Additionally, PGAM5-Bcl-xL-Keap1 interaction was identified, suggesting that PGAM5 might participate in the degradation of Bcl-xL mediated by Keap1. In summary, PGAM5 controls cardiomyocyte apoptosis induced by MIRI through regulating Keap1-mediated Bcl-xL degradation, which may supply a novel molecular target for acute myocardial infarction (AMI) therapy. Graphical abstract ᅟ.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PGAM5