MilliporeSigma
  • The relative influence of derivatization and normalization procedures on the compound-specific stable isotope analysis of nitrogen in amino acids.
The relative influence of derivatization and normalization procedures on the compound-specific stable isotope analysis of nitrogen in amino acids.
Rapid communications in mass spectrometry : RCM (2017-02-09)
Christopher T Yarnes, Julian Herszage
ABSTRACT
Within the last decade, applications of compound-specific stable isotope analysis of nitrogen (δ We examined the relative effect of three normalization procedures, (1) internal reference calibration, (2) compound-specific calibration, and (3) scale-normalization, on the accuracy and precision of δ The overall accuracy and precision of δ
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methyl chloroformate, 99%