Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Size and shape effects in β-NaGdF
Size and shape effects in β-NaGdF
Nanotechnology (2017-03-08)
Agnieszka Noculak, Artur Podhorodecki
PMID
28266926
ABSTRACT
Three sets of β-NaGdF
Product Description
544973
Sigma-Aldrich
Ytterbium(III) acetate hydrate, 99.95% trace metals basis
Pricing
325678
Sigma-Aldrich
Gadolinium(III) acetate hydrate, 99.9% trace metals basis
Pricing
325570
Sigma-Aldrich
Erbium(III) acetate hydrate, 99.9% trace metals basis
Pricing