Merck
  • Poly lactic-co-glycolic acid controlled delivery of disulfiram to target liver cancer stem-like cells.
Poly lactic-co-glycolic acid controlled delivery of disulfiram to target liver cancer stem-like cells.
Nanomedicine : nanotechnology, biology, and medicine (2016-08-16)
Zhipeng Wang, Jiao Tan, Christopher McConville, Vinodh Kannappan, Patricia Erebi Tawari, James Brown, Jin Ding, Angel L Armesilla, Juan M Irache, Qi-Bing Mei, Yuhuan Tan, Ying Liu, Wenguo Jiang, Xiu-Wu Bian, Weiguang Wang
ABSTRACT
Disulfiram (DS), an anti-alcoholism drug, shows very strong cytotoxicity in many cancer types. However its clinical application in cancer treatment is limited by the very short half-life in the bloodstream. In this study, we developed a poly lactic-co-glycolic acid (PLGA)-encapsulated DS protecting DS from the degradation in the bloodstream. The newly developed DS-PLGA was characterized. The DS-PLGA has very satisfactory encapsulation efficiency, drug-loading content and controlled release rate in vitro. PLGA encapsulation extended the half-life of DS from shorter than 2minutes to 7hours in serum. In combination with copper, DS-PLGA significantly inhibited the liver cancer stem cell population. CI-isobologram showed a remarkable synergistic cytotoxicity between DS-PLGA and 5-FU or sorafenib. It also demonstrated very promising anticancer efficacy and antimetastatic effect in liver cancer mouse model. Both DS and PLGA are FDA approved products for clinical application. Our study may lead to repositioning of DS into liver cancer treatment.
Sigma-Aldrich
Copper(II) D-gluconate, 98%
Pricing and availability is not currently available.