Merck
Inhibitory effect of
Experimental and therapeutic medicine (2017-03-30)
You-Jin Chang, Dong-Ung Lee, Da Yeong Nam, Sung Min Cho, Seungug Hong, Joo Hyun Nam, Woo Kyung Kim
ABSTRACT
In traditional Korean/Asian medicine, Salvia plebeia R.Br. (