Merck
Cubane-like tetranuclear Cu(ii) complexes bearing a Cu
Dalton transactions (Cambridge, England : 2003) (2017-01-07)
Shipra Sagar, Swaraj Sengupta, Antonio J Mota, Shyamal K Chattopadhyay, Arturo Espinosa Ferao, Eric Riviere, William Lewis, Subhendu Naskar
ABSTRACT
In the present work, two new copper complexes 3a and 3b with a Cu
Sigma-Aldrich
5-Chlorosalicylaldehyde, 98%