Merck
  • ORAI2 modulates store-operated calcium entry and T cell-mediated immunity.
ORAI2 modulates store-operated calcium entry and T cell-mediated immunity.
Nature communications (2017-03-16)
Martin Vaeth, Jun Yang, Megumi Yamashita, Isabelle Zee, Miriam Eckstein, Camille Knosp, Ulrike Kaufmann, Peter Karoly Jani, Rodrigo S Lacruz, Veit Flockerzi, Imre Kacskovics, Murali Prakriya, Stefan Feske
ABSTRACT
Store-operated Ca
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Fluorescein di(β-D-galactopyranoside)
Sigma-Aldrich
MISSION® esiRNA, targeting human ORAI2
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution