Merck
Mechanism of eukaryotic protein synthesis inhibition by brusatol.
Biochimica et biophysica acta (1981-07-27)
W Willingham, E A Stafford, S H Reynolds, S G Chaney, K H Lee, M Okano, I H Hall
ABSTRACT
The mechanism by which brusatol inhibits protein synthesis in rabbit reticulocytes has been investigated. When added to reticulocyte lysates, brusatol inhibits endogenous protein synthesis only after a lag of 2-4 min at 30 degrees C. During this period 80 S ribosomes accumulate. Brusatol is equally effective in inhibiting endogenous protein synthesis in lysates and poly(U)-directed polyphenylalanine synthesis with runoff ribosomes. In fractionated reticulocyte systems, brusatol does not inhibit formation of the ternary, 40 S, and 80 S initiation complexes, but does inhibit the reaction of puromycin with initiation complexes containing [35S]Met-tRNAf. These data suggest that brusatol inhibits the peptidyl transferase elongation reaction of protein synthesis, but can do so only after one round of protein synthesis has been completed. Thus, the mechanism of action of brusatol in the rabbit reticulocyte system is very similar to the effects previously reported for bruceantin in a yeast system.
Sigma-Aldrich
Brusatol, ≥95% (HPLC)