Merck
  • Design of isoform-selective phospholipase D inhibitors that modulate cancer cell invasiveness.
Design of isoform-selective phospholipase D inhibitors that modulate cancer cell invasiveness.
Nature chemical biology (2009-01-13)
Sarah A Scott, Paige E Selvy, Jason R Buck, Hyekyung P Cho, Tracy L Criswell, Ashley L Thomas, Michelle D Armstrong, Carlos L Arteaga, Craig W Lindsley, H Alex Brown
ABSTRACT
Phospholipase D (PLD) is an essential enzyme responsible for the production of the lipid second messenger phosphatidic acid. Phosphatidic acid participates in both G protein-coupled receptor and receptor tyrosine kinase signal transduction networks. The lack of potent and isoform-selective inhibitors has limited progress in defining the cellular roles of PLD. We used a diversity-oriented synthetic approach and developed a library of PLD inhibitors with considerable pharmacological characterization. Here we report the rigorous evaluation of that library, which contains highly potent inhibitors, including the first isoform-selective PLD inhibitors. Specific members of this series inhibit isoforms with >100-fold selectivity both in vitro and in cells. A subset of inhibitors was shown to block invasiveness in metastatic breast cancer models. These findings demonstrate the power of diversity-oriented synthesis combined with biochemical assays and mass spectrometric lipid profiling of cellular responses to develop the first isoform-selective PLD inhibitors--a new class of antimetastatic agents.
Product Description
Supelco
Diethylstilbestrol, VETRANAL®, analytical standard
Sigma-Aldrich
Diethylstilbestrol, ≥99% (HPLC)
Sigma-Aldrich
VU0155069, ≥98% (HPLC)
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Tamoxifen, ≥99%
Avanti
VU0155056, Avanti Research - A Croda Brand 857370P, powder
Sigma-Aldrich
VU0155056, ≥98% (HPLC)
Sigma-Aldrich
VU0155072, ≥98% (HPLC)