Merck
  • An IC-MS/MS Method for the Determination of 1-Hydroxyethylidene-1,1-diphosphonic Acid on Uncooked Foods Treated with Peracetic Acid-Based Sanitizers.
An IC-MS/MS Method for the Determination of 1-Hydroxyethylidene-1,1-diphosphonic Acid on Uncooked Foods Treated with Peracetic Acid-Based Sanitizers.
Chemical & pharmaceutical bulletin (2016-12-03)
Ippei Suzuki, Hiroki Kubota, Takashi Ohtsuki, Chiye Tatebe, Atsuko Tada, Takeo Yano, Hiroshi Akiyama, Kyoko Sato
ABSTRACT
A rapid, sensitive, and specific analytical method for the determination of 1-hydroxyethylidene-1,1-diphosphonic acid (HEDP) on uncooked foods after treatment with a peracetic acid-based sanitizer (PAS) was developed. The method involves simple sample preparation steps and analysis using ion chromatography (IC) coupled with tandem mass spectrometry (MS/MS). The quantification limits of HEDP on uncooked foods are 0.007 mg/kg for vegetables and fruits and 0.2 mg/kg for meats. The recovery and relative standard deviation (RSD) of HEDP analyses of uncooked foods ranged from 73.9 to 103.8% and 1.9 to 12.6%, respectively. The method's accuracy and precision were evaluated by inter-day recovery tests. The recovery for all samples ranged from 93.6 to 101.2%, and the within-laboratory repeatability and reproducibility were evaluated based on RSD values, which were less than 6.9 and 11.5%, respectively. Analyses of PAS-treated fruits and vegetables using the developed method indicated levels of HEDP ranging from 0.008 to 0.351 mg/kg. Therefore, the results of the present study suggest that the proposed method is an accurate, precise, and reliable way to determine residual HEDP levels on PAS-treated uncooked foods.
Sigma-Aldrich
Zoledronic acid monohydrate, ≥98% (HPLC)
Sigma-Aldrich
Etidronic acid, 60% aqueous solution