Merck
  • Fluorine-19 NMR and computational quantification of isoflurane binding to the voltage-gated sodium channel NaChBac.
Fluorine-19 NMR and computational quantification of isoflurane binding to the voltage-gated sodium channel NaChBac.
Proceedings of the National Academy of Sciences of the United States of America (2016-11-20)
Monica N Kinde, Vasyl Bondarenko, Daniele Granata, Weiming Bu, Kimberly C Grasty, Patrick J Loll, Vincenzo Carnevale, Michael L Klein, Roderic G Eckenhoff, Pei Tang, Yan Xu
ABSTRACT
Voltage-gated sodium channels (NaV) play an important role in general anesthesia. Electrophysiology measurements suggest that volatile anesthetics such as isoflurane inhibit NaV by stabilizing the inactivated state or altering the inactivation kinetics. Recent computational studies suggested the existence of multiple isoflurane binding sites in NaV, but experimental binding data are lacking. Here we use site-directed placement of 19F probes in NMR experiments to quantify isoflurane binding to the bacterial voltage-gated sodium channel NaChBac. 19F probes were introduced individually to S129 and L150 near the S4-S5 linker, L179 and S208 at the extracellular surface, T189 in the ion selectivity filter, and all phenylalanine residues. Quantitative analyses of 19F NMR saturation transfer difference (STD) spectroscopy showed a strong interaction of isoflurane with S129, T189, and S208; relatively weakly with L150; and almost undetectable with L179 and phenylalanine residues. An orientation preference was observed for isoflurane bound to T189 and S208, but not to S129 and L150. We conclude that isoflurane inhibits NaChBac by two distinct mechanisms: (i) as a channel blocker at the base of the selectivity filter, and (ii) as a modulator to restrict the pivot motion at the S4-S5 linker and at a critical hinge that controls the gating and inactivation motion of S6.
Sigma-Aldrich
m-Fluoro-DL-phenylalanine