Merck
  • Formulation of yeast-leavened bread with reduced salt content by using a Lactobacillus plantarum fermentation product.
Formulation of yeast-leavened bread with reduced salt content by using a Lactobacillus plantarum fermentation product.
Food chemistry (2016-12-17)
Francesca Valerio, Amalia Conte, Mariaelena Di Biase, Veronica M T Lattanzio, S Lisa Lonigro, Lucia Padalino, Erica Pontonio, Paola Lavermicocca
ABSTRACT
A Lactobacillus plantarum fermentation product (Bio21B), obtained after strain growth (14h) in a wheat flour-based medium, was applied in the bread-making process as taste enhancer, in order to obtain a yeast-leavened bread with reduced salt content (20% and 50%) with respect to a reference bread (REF) not containing the fermentation product. Sensory analysis indicated that the Bio21B bread with salt reduced by 50% had a pleasant taste similar to the salt-containing bread (REF). l-Glutamate and total free amino acid content did not differ between REF and Bio21B breads, while the acids lactic, acetic, phenyllactic, 4-OH-phenyllactic and indole-3-lactic were present only in Bio21B breads. Moreover, the presence of several umami (uridine monophosphate, inosine monophosphate, adenosine, and guanosine) and kokumi (γ-l-glutamyl-l-valine) taste-related molecules was ascertained both in REF and in Bio21B breads. Therefore, a possible role of the acidic molecules in compensating the negative perception of salt reduction can be hypothesized.