Merck
  • PML isoform II plays a critical role in nuclear lipid droplet formation.
PML isoform II plays a critical role in nuclear lipid droplet formation.
The Journal of cell biology (2016-01-06)
Yuki Ohsaki, Takeshi Kawai, Yukichika Yoshikawa, Jinglei Cheng, Eija Jokitalo, Toyoshi Fujimoto
ABSTRACT
Lipid droplets (LDs) in the nucleus of hepatocyte-derived cell lines were found to be associated with premyelocytic leukemia (PML) nuclear bodies (NBs) and type I nucleoplasmic reticulum (NR) or the extension of the inner nuclear membrane. Knockdown of PML isoform II (PML-II) caused a significant decrease in both nuclear LDs and type I NR, whereas overexpression of PML-II increased both. Notably, these effects were evident only in limited types of cells, in which a moderate number of nuclear LDs exist intrinsically, and PML-II was targeted not only at PML NBs, but also at the nuclear envelope, excluding lamins and SUN proteins. Knockdown of SUN proteins induced a significant increase in the type I NR and nuclear LDs, but these effects were cancelled by simultaneous knockdown of PML-II. Nuclear LDs harbored diacylglycerol O-acyltransferase 2 and CTP:phosphocholine cytidylyltransferase α and incorporated newly synthesized lipid esters. These results corroborated that PML-II plays a critical role in generating nuclear LDs in specific cell types.
Product Description
Avanti
18:1-C11 TopFluor DG, Avanti Research - A Croda Brand 810297P, powder
Avanti
C4 TopFluor MG, Avanti Research - A Croda Brand 810293P, powder
Avanti
C11 TopFluor MG, Avanti Research - A Croda Brand 810296P, powder
Avanti
18:1-6:0 DNP-C11 TopFluor TG, Avanti Research - A Croda Brand 810272P, powder
Avanti
18:1-C11 TopFluor-C11 TopFluor TG, Avanti Research - A Croda Brand 810271P, powder
Avanti
18:1-18:1-C11 TopFluor TG, Avanti Research - A Croda Brand 810298C
Sigma-Aldrich
Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse, clone SC-35, ascites fluid