Merck
  • Extrusion of mitochondrial contents from lipopolysaccharide-stimulated cells: Involvement of autophagy.
Extrusion of mitochondrial contents from lipopolysaccharide-stimulated cells: Involvement of autophagy.
Autophagy (2015-06-24)
Kana Unuma, Toshihiko Aki, Takeshi Funakoshi, Kyoko Hashimoto, Koichi Uemura
ABSTRACT
Sepsis/endotoxemia is elicited by the circulatory distribution of pathogens/endotoxins into whole bodies, and causes profound effects on human health by causing inflammation in multiple organs. Mitochondrial damage is one of the characteristics of the cellular degeneration observed during sepsis/endotoxemia. Elimination of damaged mitochondria through the autophagy-lysosome system has been reported in the liver, indicating that autophagy should play an important role in liver homeostasis during sepsis/endotoxemia. An increased appearance of mitochondrial DNA and proteins in the plasma is another feature of sepsis/endotoxemia, suggesting that damaged mitochondria are not only eliminated within the cells, but also extruded through currently unknown mechanisms. Here we provide evidence for the secretion of mitochondrial proteins and DNA from lipopolysaccharide (LPS)-stimulated rat hepatocytes as well as mouse embryonic fibroblasts (MEFs). The secretion of mitochondrial contents is accompanied by the secretion of proteins that reside in the lumenal space of autolysosomes (LC3-II and CTSD/cathepsin D), but not by a lysosomal membrane protein (LAMP1). The pharmacological inhibition of autophagy by 3MA blocks the secretion of mitochondrial constituents from LPS-stimulated hepatocytes. LPS also stimulates the secretion of mitochondrial as well as autolysosomal lumenal proteins from wild-type (Atg5(+/+)) MEFs, but not from atg5(-/-) MEFs. Furthermore, we show that direct exposure of purified mitochondria activates polymorphonuclear leukocytes (PMNs), as evident by the induction of IL1B/interlekin-1β, a pro-inflammatory cytokine. Taken together, the data suggest the active extrusion of mitochondrial contents, which provoke an inflammatory response of immune cells, through the exocytosis of autolysosomes by cells stimulated with LPS.
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Caspase Inhibitor I, Z-VAD-FMK, CAS 187389-52-2, is a cell-permeable, irreversible, pan-caspase inhibitor.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Dynasore hydrate
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution