Merck
  • In vitro depletion of tissue-derived dendritic cells by CMRF-44 antibody and alemtuzumab: implications for the control of Graft-versus-host disease.
In vitro depletion of tissue-derived dendritic cells by CMRF-44 antibody and alemtuzumab: implications for the control of Graft-versus-host disease.
Transplantation (2005-03-24)
Matthew P Collin, David Munster, Georgina Clark, Xiao-Nong Wang, Anne M Dickinson, Derek N Hart
ABSTRACT
Graft-versus-host disease (GvHD), a life-threatening complication of bone marrow transplantation, is initiated by donor T cells reacting to recipient dendritic cells (DC). GvHD can be controlled by attenuating donor T cells, but few strategies exist to target DC, particularly resident tissue DC, despite recent evidence of their importance. In this report, CMRF-44, a mouse monoclonal IgM reactive to human DC, is tested against human Langerhans cells (LC) in vitro. CMRF-44 antigen is expressed at low level on fresh LC but is up-regulated 40-60-fold during migration. CMRF-44 and complement kill more than 97% of migratory LC in vitro and inhibit allostimulation by LC up to 95%. In comparison, alemtuzumab, which binds CD52, reacts weakly with primary LC and fails to induce significant lysis with complement (less than 5%). These results highlight the potential of new therapeutic antibodies active against tissue DC to control graft-versus-host reactions.
Sigma-Aldrich
IgM, Kappa from murine myeloma, clone TEPC 183, purified immunoglobulin, buffered aqueous solution