Merck
TAp63gamma is required for the late stages of myogenesis.
Cell cycle (Georgetown, Tex.) (2015-03-20)
S Cefalù, A M Lena, B Vojtesek, A Musarò, A Rossi, G Melino, E Candi
ABSTRACT
p53 family members, p63 and p73, play a role in controlling early stage of myogenic differentiation. We demonstrated that TAp63gamma, unlike the other p53 family members, is markedly up-regulated during myogenic differentiation in murine C2C7 cell line. We also found that myotubes formation was inhibited upon TAp63gamma knock-down, as also indicated by atrophyic myotubes and reduction of myoblasts fusion index. Analysis of TAp63gamma-dependend transcripts identified several target genes involved in skeletal muscle contractility energy metabolism, myogenesis and skeletal muscle autocrine signaling. These results indicate that TAp63gamma is a late marker of myogenic differentiation and, by controlling different sub-sets of target genes, it possibly contributes to muscle growth, remodeling, functional differentiation and tissue homeostasis.
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid