Merck
  • Candida albicans Dbf4-dependent Cdc7 kinase plays a novel role in the inhibition of hyphal development.
Candida albicans Dbf4-dependent Cdc7 kinase plays a novel role in the inhibition of hyphal development.
Scientific reports (2016-09-21)
Wei-Chung Lai, Tschen-Wei Chang, Chang Hao Wu, Shu-Ya Yang, Tai-Lin Lee, Wan Chen Li, Ting Chien, Yu-Che Cheng, Jia-Ching Shieh
ABSTRACT
Candida albicans is an opportunistic human fungal pathogen. The ability to switch among multiple cellular forms is key to its pathogenesis. The Dbf4-dependent protein kinase gene CDC7 is conserved due to its role in initiating DNA replication. Because a C. albicans Cdc7 (Cacdc7) homozygous null was not viable, we generated a C. albicans strain with a deleted C. albicans CDC7 (CaCDC7) allele and an expression-repressible allele. Surprisingly, cells of the strain grew as hyphae under the repressed conditions. The in vitro kinase assays confirmed that CaCdc7 (K232) and CaCdc7 (T437) are critical for catalytic and phosphoacceptor of activation activity, respectively. C. albicans cells formed hyphae when expressing either the catalytically inactive CaCdc7 (K232R) or the phosphoacceptor-deficient CaCdc7 (T437A). While CaCdc7 interacted with CaDbf4, cells of the strain in which CaCDC7 was repressed were not rescued by constitutively expressing C. albicans DBF4 or vice versa. We conclude that CaDBF4-dependent CaCDC7 is an essential gene suppressing the hyphal development.
Sigma-Aldrich
Anti-Histone Antibody, clone H11-4, clone H11-4, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H1 Antibody, clone 12D11, culture supernatant, clone 12D11, from mouse