Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Novel selective PDE4 inhibitors. 2. Synthesis and structure-activity relationships of 4-aryl-substituted cis-tetra- and cis-hexahydrophthalazinones.
Novel selective PDE4 inhibitors. 2. Synthesis and structure-activity relationships of 4-aryl-substituted cis-tetra- and cis-hexahydrophthalazinones.
Journal of medicinal chemistry (2001-07-27)
M Van der Mey, A Hatzelmann, G P Van Klink, I J Van der Laan, G J Sterk, U Thibaut, W R Ulrich, H Timmerman
ABSTRACT
A series of 4-aryl-substituted cis-4a,5,8,8a-tetra- and cis-4a,5,6,7,8,8a-hexahydro-2H-phthalazin-1-ones with high inhibitory activity toward cAMP-specific phosphodiesterase (PDE4) was synthesized. To study structure-activity relationships various substituents were introduced to the 2-, 3-, and 4-positions of the 4-phenyl ring. Substitution at the 4-position of the phenyl ring was restricted to a methoxy group, probably due to unfavorable steric interactions of larger groups with the binding site. The introduction of many alkoxy substituents including distinct ring systems and functional groups was allowed to the 3-position. It was found that in general the cis-4a,5,8,8a-tetrahydro-2H-phthalazin-1-ones are more potent than their hexahydrophthalic counterparts, the best activity residing in (4-imidazol-1-yl-phenoxy)butoxy analogue 16o (pIC(50) = 9.7).