Merck
  • Total synthesis of (-)-martinellic acid via radical addition-cyclization-elimination reaction.
Total synthesis of (-)-martinellic acid via radical addition-cyclization-elimination reaction.
The Journal of organic chemistry (2008-05-14)
Atsushi Shirai, Okiko Miyata, Norimitsu Tohnai, Mikiji Miyata, David J Procter, David Sucunza, Takeaki Naito
ABSTRACT
The asymmetric total synthesis of martinellic acid, the first pyrrolo[3,2-c]quinoline alkaloid found in nature, is described. Three key steps in our synthesis of (-)-martinellic acid are the Bu(3)SnH-promoted radical addition-cyclization-elimination (RACE) reaction of an oxime ether with an alpha,beta-unsaturated ester to generate the pyrrolo[3,2-c]quinoline core, a chemoselective lactam carbonyl reduction, and guanidinylation under Mitsunobu reaction conditions. The key radical cyclization has also been investigated by using SmI(2). (-)-Martinellic acid was synthesized from commercially available methyl 4-bromo-3-methylbenzoate in fewer steps than previous syntheses and in an improved overall yield.
Sigma-Aldrich
Methyl 4-bromo-3-methylbenzoate, 95%