Merck
  • Novel role for the midbody in primary ciliogenesis by polarized epithelial cells.
Novel role for the midbody in primary ciliogenesis by polarized epithelial cells.
The Journal of cell biology (2016-07-28)
Miguel Bernabé-Rubio, Germán Andrés, Javier Casares-Arias, Jaime Fernández-Barrera, Laura Rangel, Natalia Reglero-Real, David C Gershlick, José J Fernández, Jaime Millán, Isabel Correas, David G Miguez, Miguel A Alonso
ABSTRACT
The primary cilium is a membrane protrusion that is crucial for vertebrate tissue homeostasis and development. Here, we investigated the uncharacterized process of primary ciliogenesis in polarized epithelial cells. We show that after cytokinesis, the midbody is inherited by one of the daughter cells as a remnant that initially locates peripherally at the apical surface of one of the daughter cells. The remnant then moves along the apical surface and, once proximal to the centrosome at the center of the apical surface, enables cilium formation. The physical removal of the remnant greatly impairs ciliogenesis. We developed a probabilistic cell population-based model that reproduces the experimental data. In addition, our model explains, solely in terms of cell area constraints, the various observed transitions of the midbody, the beginning of ciliogenesis, and the accumulation of ciliated cells. Our findings reveal a biological mechanism that links the three microtubule-based organelles-the midbody, the centrosome, and the cilium-in the same cellular process.
Sigma-Aldrich
Anti-IFT20 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Rab8 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution