Merck
  • DNA methylation reactivates GAD1 expression in cancer by preventing CTCF-mediated polycomb repressive complex 2 recruitment.
DNA methylation reactivates GAD1 expression in cancer by preventing CTCF-mediated polycomb repressive complex 2 recruitment.
Oncogene (2015-11-10)
H Yan, G Tang, H Wang, L Hao, T He, X Sun, A H Ting, A Deng, S Sun
ABSTRACT
Levels of γ-aminobutyric acid (GABA) and glutamic acid decarboxylase 1 (GAD1), the enzyme that synthesizes GABA, are significantly increased in neoplastic tissues. However, the mechanism underlying this increase remains elusive. Instead of silencing gene transcription, we showed that the GAD1 promoter was hypermethylated in both colon and liver cancer cells, leading to the production of high levels of GAD1. GAD1 is a target gene that is silenced by H3K27me3. The key locus responsible for GAD1 reactivation was mapped to a DNA methylation-sensitive CTCF-binding site (CTCF-BS3) within the third intron of GAD1. Chromosome configuration capture (3C) analysis indicated that an intrachromosomal loop was formed by CTCF self-dimerisation in normal cells (CTCF binds to both unmethylated CTCF-BS3 and CTCF-BS2). The CTCF dimer then interacted with suppressor of zeste 12 homologue (SUZ12), which is a domain of Polycomb repressive complex 2 (PRC2), promoting the methylation of H3K27 and the silencing of GAD1 expression. This silencing was shown to be inhibited by DNA methylation in cancer cells. These findings strongly suggest that GAD1 is reactivated by DNA methylation, which provided a model for DNA methylation and the active orchestration of oncogenic gene expression by CTCF in cancer cells.
Sigma-Aldrich
ChIPAb+ CTCF - ChIP Validated Antibody and Primer Set, clone 1.2.1.5.4, from mouse
Sigma-Aldrich
Monoclonal Anti-Glutamic Acid Decarboxylase 67 (GAD67) antibody produced in mouse, clone K-87, purified antibody