Merck
  • Structural insights into Ca(2+)-activated long-range allosteric channel gating of RyR1.
Structural insights into Ca(2+)-activated long-range allosteric channel gating of RyR1.
Cell research (2016-08-31)
Risheng Wei, Xue Wang, Yan Zhang, Saptarshi Mukherjee, Lei Zhang, Qiang Chen, Xinrui Huang, Shan Jing, Congcong Liu, Shuang Li, Guangyu Wang, Yaofang Xu, Sujie Zhu, Alan J Williams, Fei Sun, Chang-Cheng Yin
ABSTRACT
Ryanodine receptors (RyRs) are a class of giant ion channels with molecular mass over 2.2 mega-Daltons. These channels mediate calcium signaling in a variety of cells. Since more than 80% of the RyR protein is folded into the cytoplasmic assembly and the remaining residues form the transmembrane domain, it has been hypothesized that the activation and regulation of RyR channels occur through an as yet uncharacterized long-range allosteric mechanism. Here we report the characterization of a Ca(2+)-activated open-state RyR1 structure by cryo-electron microscopy. The structure has an overall resolution of 4.9 Å and a resolution of 4.2 Å for the core region. In comparison with the previously determined apo/closed-state structure, we observed long-range allosteric gating of the channel upon Ca(2+) activation. In-depth structural analyses elucidated a novel channel-gating mechanism and a novel ion selectivity mechanism of RyR1. Our work not only provides structural insights into the molecular mechanisms of channel gating and regulation of RyRs, but also sheds light on structural basis for channel-gating and ion selectivity mechanisms for the six-transmembrane-helix cation channel family.
Sigma-Aldrich
Soy Phospholipids, mixture of phospholipids
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Pricing and availability is not currently available.