Merck
  • The long noncoding RNA ASNR regulates degradation of Bcl-2 mRNA through its interaction with AUF1.
The long noncoding RNA ASNR regulates degradation of Bcl-2 mRNA through its interaction with AUF1.
Scientific reports (2016-09-01)
Jiahui Chen, Lihui Liu, Guifeng Wei, Wei Wu, Huaxia Luo, Jiao Yuan, Jianjun Luo, Runsheng Chen
ABSTRACT
The identification and characterization of long non-coding RNAs (lncRNAs) in diverse biological processes has recently developed rapidly. The large amounts of non-coding RNAs scale consistent with developmental complexity in eukaryotes, indicating that most of these transcripts may have functions in the regulation of biological processes and disorder in the organisms. In particular, Understanding of the overall biological significance of lncRNAs in cancers still remains limited. Here, we found a nuclear-retained lncRNA, termed Lnc_ASNR (apoptosis suppressing-noncoding RNA), which serves as a repressor of apoptosis. Lnc_ASNR was discovered in a set of microarray data derived from four kinds of tumor and adjacent normal tissue samples, and displayed significant up-regulation in the tumor tissues. Using an RNA-pull down assay, we found that Lnc_ASNR interacted with the protein ARE/poly (U)-binding/degradation factor 1(AUF1), which is reported to promote rapid degradation of the Bcl-2 mRNA, an inhibitor of apoptosis. Lnc_ASNR binds to AUFI in nucleus, decreasing the cytoplasmic proportion of AUF1 which targets the B-cell lymphoma-2 (Bcl-2) mRNA. Taken together, the overall effect of Lnc_ASNR expression is thus a decrease in cell apoptosis indicating that Lnc_ASNR may play a vital role in tumorigenesis and carcinogenesis.
Sigma-Aldrich
RIPAb+ AUF1 - RIP Validated Antibody and Primer Set, from rabbit