Merck
Structural basis for acyl-group discrimination by human Gcn5L2.
Acta crystallographica. Section D, Structural biology (2016-07-06)
Alison E Ringel, Cynthia Wolberger
ABSTRACT
Gcn5 is a conserved acetyltransferase that regulates transcription by acetylating the N-terminal tails of histones. Motivated by recent studies identifying a chemically diverse array of lysine acyl modifications in vivo, the acyl-chain specificity of the acetyltransferase human Gcn5 (Gcn5L2) was examined. Whereas Gcn5L2 robustly catalyzes lysine acetylation, the acyltransferase activity of Gcn5L2 becomes progressively weaker with increasing acyl-chain length. To understand how Gcn5 discriminates between different acyl-CoA molecules, structures of the catalytic domain of human Gcn5L2 bound to propionyl-CoA and butyryl-CoA were determined. Although the active site of Gcn5L2 can accommodate propionyl-CoA and butyryl-CoA without major structural rearrangements, butyryl-CoA adopts a conformation incompatible with catalysis that obstructs the path of the incoming lysine residue and acts as a competitive inhibitor of Gcn5L2 versus acetyl-CoA. These structures demonstrate how Gcn5L2 discriminates between acyl-chain donors and explain why Gcn5L2 has weak activity for acyl moieties that are larger than an acetyl group.
Sigma-Aldrich
n-Propionyl coenzyme A lithium salt, ≥85%
Sigma-Aldrich
Butyryl coenzyme A lithium salt hydrate, ≥90%
Sigma-Aldrich
Acetyl coenzyme A lithium salt, ≥93% (HPLC)