Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Proteolytic disassembly of peptide-mediated graphene oxide assemblies for turn-on fluorescence sensing of proteases.
Proteolytic disassembly of peptide-mediated graphene oxide assemblies for turn-on fluorescence sensing of proteases.
Nanoscale (2016-06-09)
Jin-Kyoung Yang, Seon-Yeong Kwak, Su-Ji Jeon, Eunjin Lee, Jong-Min Ju, Hye-In Kim, Yoon-Sik Lee, Jong-Ho Kim
ABSTRACT
Molecule-induced assembly of nanomaterials can alter their unique chemical and physical properties, which can be a promising approach for sensing. Herein, we demonstrate an optical 'turn-on' biosensor for the detection of matrix metalloproteinase-2 (MMP-2), fabricated by means of a peptide-induced assembly of fluorescent graphene oxide (GO). Functionalization of GO with a peptide substrate for MMP-2 bearing a thiol group leads to its self-assembly via disulfide bonding, accompanied by self-quenching of GO's strong fluorescence. This peptide-induced GO assembly is then disassembled by proteolytic cleavage in the presence of MMP-2, thereby restoring the level of self-quenched GO fluorescence. With this approach, we are able to detect MMP-2 and to investigate the kinetic parameters of MMP-2 activity. The GO-peptide assembly is successfully applied to the selective and sensitive detection of MMP-2 secreted by living cells, human hepatocytes HepG2, at a concentration of 2 ng mL(-1).
Sigma-Aldrich
Prinomastat hydrochloride, ≥95% (HPLC)