Merck
Co-expression of anoctamins in cilia of olfactory sensory neurons.
Chemical senses (2014-12-17)
Bastian Henkel, Daniela R Drose, Tobias Ackels, Sonja Oberland, Marc Spehr, Eva M Neuhaus
ABSTRACT
Vertebrates can sense and identify a vast array of chemical cues. The molecular machinery involved in chemodetection and transduction is expressed within the cilia of olfactory sensory neurons. Currently, there is only limited information available on the distribution and density of individual signaling components within the ciliary compartment. Using super-resolution microscopy, we show here that cyclic-nucleotide-gated channels and calcium-activated chloride channels of the anoctamin family are localized to discrete microdomains in the ciliary membrane. In addition to ANO2, a second anoctamin, ANO6, also localizes to ciliary microdomains. This observation, together with the fact that ANO6 and ANO2 co-localize, indicates a role for ANO6 in olfactory signaling. We show that both ANO2 and ANO6 can form heteromultimers and that this heteromerization alters the recombinant channels' physiological properties. Thus, we provide evidence for interaction of ANO2 and ANO6 in olfactory cilia, with possible physiological relevance for olfactory signaling.
Sigma-Aldrich
Anti-Renilla Luciferase Antibody, clone 5B11.2, ascites fluid, clone 5B11.2, Chemicon®
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, biotin-SP conjugate, Species Adsorbed, 1 mg/mL, Chemicon®