Merck
  • A facile one-pot preparation of alkyl aminoaryl sulfides for the synthesis of GW7647 as an agonist of peroxisome proliferator-activated receptor alpha.
A facile one-pot preparation of alkyl aminoaryl sulfides for the synthesis of GW7647 as an agonist of peroxisome proliferator-activated receptor alpha.
The Journal of organic chemistry (2006-07-15)
Jungyeob Ham, Sung Jin Cho, Jaeyoung Ko, Jungwook Chin, Heonjoong Kang
ABSTRACT
We have developed two simple and high yielding one-pot syntheses of alkyl aminoaryl sulfides containing a series of four-steps: in situ protection of the free amine by reaction with a Grignard reagent, halogen-lithium exchange, sulfur insertion, and a substitution reaction with various electrophiles. Through this protocol, we have successfully synthesized tert-butyl-2-[4-(2-aminoethyl)phenylsulfanyl]-2-methylpropanoate, a key intermediate for the synthesis of GW7647 and GW9578 (ureido-TiBAs), in 92% yield. Furthermore, we were able to improve the overall yield of GW7647 to 66%, 3 times the yield previously reported.