Merck
Tandem isomerization/telomerization of long chain dienes.
Frontiers in chemistry (2014-07-02)
Laura Torrente-Murciano, David J Nielsen, Kingsley J Cavell, Alexei A Lapkin
ABSTRACT
The first example of a tandem reaction involving double-bond migration in combination with telomerization is reported. Homogeneous and heterogeneous Ru catalysts were employed as isomerization catalysts, and telomerization was realized using a homogeneous Pd(0) precursor complex with a N-heterocyclic carbene (IMes) ligand. Overall conversions approaching 60% were achieved with the best selectivity to telomerization products of 91% attained at 11% conversion. Conversion was markedly higher in the presence of longer-chain alcohol (1-butanol) as the nucleophile (telogen).
Sigma-Aldrich
1,3-Hexadiene, mixture of cis and trans, 95%