Merck
  • Emerging concepts in GPCR function--the influence of cell phenotype on GPCR pharmacology.
Emerging concepts in GPCR function--the influence of cell phenotype on GPCR pharmacology.
Proceedings of the Western Pharmacology Society (2006-01-19)
Richard M Eglen
ABSTRACT
G protein coupled receptors (GPCRs) are an important class of ligand-activated proteins that regulate cellular metabolism. It is now appears that GPCR function is highly dependent upon the cellular environment in which it is expressed. Indeed, the cell phenotype influences several sequelae of GPCR activation, including GPCR oligomerization, G protein coupling, spatial organization of signaling proteins and inactivation and internalization processes. The cell phenotype, consequently, modulates both the affinity and efficacy of endogenous agonists and synthetic compounds. Emerging data suggests a complex function of GPCR signaling resulting in tissue specific responses, even those emanating from the same GPCR subtype. This short review describes some phenotypic influences on GPCR function and their implications for the use of cell-based assays used to identify and characterize GPCR ligands.
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, G-Protein Coupled Receptors
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Bacterial Glycerol Stock, G-Protein Coupled Receptors