Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Vitamin E analogues as inducers of apoptosis: structure-function relation.
Vitamin E analogues as inducers of apoptosis: structure-function relation.
British journal of cancer (2003-06-12)
M Birringer, J H EyTina, B A Salvatore, J Neuzil
ABSTRACT
Recent results show that alpha-tocopheryl succinate (alpha-TOS) is a proapoptotic agent with antineoplastic activity. As modifications of the vitamin E (VE) molecule may affect its apoptogenic activity, we tested a number of newly synthesised VE analogues using malignant cell lines. Analogues of alpha-TOS with lower number of methyl substitutions on the aromatic ring were less active than alpha-TOS. Replacement of the succinyl group with a maleyl group greatly enhanced the activity, while it was lower for the glutaryl esters. Methylation of the free succinyl carboxyl group on alpha-TOS and delta-TOS completely prevented the apoptogenic activity of the parent compounds. Both Trolox and its succinylated derivative were inactive. alpha-tocotrienol (alpha-T3 H) failed to induce apoptosis, while gamma-T3 H was apoptogenic, and more so when succinylated. Shortening the aliphatic side chain of gamma-T3 by one isoprenyl unit increased its activity. Neither phytyl nor oleyl succinate caused apoptosis. These findings show that modifications of different functional moieties of the VE molecule can enhance apoptogenic activity. It is hoped that these observations will lead to the synthesis of analogues with even higher apoptogenic and, consequently, antineoplastic efficacy.