Merck
  • CDK8-mediated STAT1-S727 phosphorylation restrains NK cell cytotoxicity and tumor surveillance.
CDK8-mediated STAT1-S727 phosphorylation restrains NK cell cytotoxicity and tumor surveillance.
Cell reports (2013-08-13)
Eva Maria Putz, Dagmar Gotthardt, Gregor Hoermann, Agnes Csiszar, Silvia Wirth, Angelika Berger, Elisabeth Straka, Doris Rigler, Barbara Wallner, Amanda M Jamieson, Winfried F Pickl, Eva Maria Zebedin-Brandl, Mathias Müller, Thomas Decker, Veronika Sexl
ABSTRACT
The transcription factor STAT1 is important in natural killer (NK) cells, which provide immediate defense against tumor and virally infected cells. We show that mutation of a single phosphorylation site (Stat1-S727A) enhances NK cell cytotoxicity against a range of tumor cells, accompanied by increased expression of perforin and granzyme B. Stat1-S727A mice display significantly delayed disease onset in NK cell-surveilled tumor models including melanoma, leukemia, and metastasizing breast cancer. Constitutive phosphorylation of S727 depends on cyclin-dependent kinase 8 (CDK8). Inhibition of CDK8-mediated STAT1-S727 phosphorylation may thus represent a therapeutic strategy for stimulating NK cell-mediated tumor surveillance.
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
Roscovitine, A potent, reversible, and selective inhibitor of Cdks that exhibits about 10-fold greater efficacy towards p34-cdk1 and p33-cdk2 and 20-fold greater efficacy towards p33-cdk5 relative to Olomoucine.
Sigma-Aldrich
Roscovitine, ≥98% (TLC)
Sigma-Aldrich
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid