Merck
  • Garlic attenuates cardiac oxidative stress via activation of PI3K/AKT/Nrf2-Keap1 pathway in fructose-fed diabetic rat.
Garlic attenuates cardiac oxidative stress via activation of PI3K/AKT/Nrf2-Keap1 pathway in fructose-fed diabetic rat.
PloS one (2014-05-07)
Raju Padiya, Debabrata Chowdhury, Roshan Borkar, R Srinivas, Manika Pal Bhadra, Sanjay K Banerjee
ABSTRACT
Cardiovascular complication due to diabetes has remained a major cause of death. There is an urgent need to intervene the cardiac complications in diabetes by nutritional or pharmacological agents. Thus the present study was designed to find out the effectiveness of garlic on cardiac complications in insulin-resistant diabetic rats. SD rats were fed high fructose (65%) diet alone or along with raw garlic homogenate (250 mg/kg/day) or nutrient-matched (65% corn starch) control diet for 8 weeks. Fructose-fed diabetic rats showed cardiac hypertrophy, increased NFkB activity and increased oxidative stress. Administration of garlic significantly decreased (p<0.05) cardiac hypertrophy, NFkB activity and oxidative stress. Although we did not observe any changes in myocardial catalase, GSH and GPx in diabetic heart, garlic administration showed significant (p<0.05) increase in all three antioxidant/enzymes levels. Increased endogenous antioxidant enzymes and gene expression in garlic treated diabetic heart are associated with higher protein expression of Nrf2. Increased myocardial H2S levels, activation of PI3K/Akt pathway and decreased Keap levels in fructose-fed heart after garlic administration might be responsible for higher Nrf2 levels. Our study demonstrates that raw garlic homogenate is effective in reducing cardiac hypertrophy and fructose-induced myocardial oxidative stress through PI3K/AKT/Nrf2-Keap1 dependent pathway.
Sigma-Aldrich
Anti-Lamin A Antibody, CT, a.a. 598-611, clone 133A2, clone 133A2, Chemicon®, from mouse