Merck
  • Fractalkine receptor is expressed in mature ovarian teratomas and required for epidermal lineage differentiation.
Fractalkine receptor is expressed in mature ovarian teratomas and required for epidermal lineage differentiation.
Journal of ovarian research (2013-08-21)
Lisa Rooper, Hilal Gurler, Andre A Kajdacsy-Balla, Maria V Barbolina
ABSTRACT
The goal of this study was to determine a predominant cell type expressing fractalkine receptor (CX3CR1) in mature ovarian teratomas and to establish functional significance of its expression in cell differentiation. Specimens of ovarian teratoma and human fetal tissues were analyzed by immunohistochemistry for CX3CR1expression. Ovarian teratocarcinoma cell line PA-1 was used as a model for cell differentiation. We found that the majority of the specimens contained CX3CR1-positive cells of epidermal lineage. Skin keratinocytes in fetal tissues were also CX3CR1- positive. PA-1 cells with downregulated CX3CR1 failed to express a skin keratinocyte marker cytokeratin 14 when cultured on Matrigel in the presence of a morphogen, bone morphogenic protein 4 (BMP-4), as compared to those expressing scrambled shRNA. Here we demonstrate that CX3CR1 is expressed in both normally (fetal skin) and abnormally (ovarian teratoma) differentiated keratinocytes and is required for cell differentiation into epidermal lineage.
Sigma-Aldrich
MISSION® esiRNA, targeting human CX3CR1
Pricing and availability is not currently available.