Merck
  • In Vitro Influence of Mycophenolic Acid on Selected Parameters of Stimulated Peripheral Canine Lymphocytes.
In Vitro Influence of Mycophenolic Acid on Selected Parameters of Stimulated Peripheral Canine Lymphocytes.
PloS one (2016-05-04)
Maciej Guzera, Lidia Szulc-Dąbrowska, Anna Cywińska, Joy Archer, Anna Winnicka
ABSTRACT
Mycophenolic acid (MPA) is an active metabolite of mycophenolate mofetil, a new immunosuppressive drug effective in the treatment of canine autoimmune diseases. The impact of MPA on immunity is ambiguous and its influence on the canine immune system is unknown. The aim of the study was to determine markers of changes in stimulated peripheral canine lymphocytes after treatment with MPA in vitro. Twenty nine healthy dogs were studied. Phenotypic and functional analysis of lymphocytes was performed on peripheral blood mononuclear cells cultured with mitogens and different MPA concentrations- 1 μM (10(-3) mol/m(3)), 10 μM or 100 μM. Apoptotic cells were detected by Annexin V and 7-aminoactinomycin D (7-AAD). The expression of antigens (CD3, CD4, CD8, CD21, CD25, forkhead box P3 [FoxP3] and proliferating cell nuclear antigen [PCNA]) was assessed with monoclonal antibodies. The proliferation indices were analyzed in carboxyfluorescein diacetate succinimidyl ester (CFSE)-labeled cells. All analyses were performed using flow cytometry. The influence of MPA on apoptosis was dependent on the mechanism of cell activation and MPA concentration. MPA caused a decrease in the expression of lymphocyte surface antigens, CD3, CD8 and CD25. Its impact on the expression of CD4 and CD21 was negligible. Its negative influence on the expression of FoxP3 was dependent on cell stimulation. MPA inhibited lymphocyte proliferation. In conclusion, MPA inhibited the activity of stimulated canine lymphocytes by blocking lymphocyte activation and proliferation. The influence of MPA on the development of immune tolerance-expansion of Treg cells and lymphocyte apoptosis-was ambiguous and was dependent on the mechanism of cellular activation. The concentration that MPA reaches in the blood may lead to inhibition of the functions of the canine immune system. The applied panel of markers can be used for evaluation of the effects of immunosuppressive compounds in the dog.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Penicillin − Streptomycin − Neomycin Solution Stabilized, formulated to contain ~5,000 units penicillin, 5 mg streptomycin and 10 mg neomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma