Merck
An efficient synthesis of an alphavbeta3 antagonist.
The Journal of organic chemistry (2004-04-03)
Nobuyoshi Yasuda, Yi Hsiao, Mark S Jensen, Nelo R Rivera, Chunhua Yang, Kenneth M Wells, James Yau, Michael Palucki, Lushi Tan, Peter G Dormer, Ralph P Volante, David L Hughes, Paul J Reider
ABSTRACT
A practical preparation of an alpha(v)beta(3) antagonist is reported. The antagonist consists of three key components, a tetrahydronaphthyridine moiety, a beta-alanine moiety, and a central imidazolidone moiety. The tetrahydronaphthyridine component was prepared using two different methods, both of which relied on variations of the Friedländer reaction to establish the desired regiochemistry. The beta-alanine component was prepared using Davies' asymmetric 1,4-addition methodology as the key stereo-defining step. The central imidazolidone portion was created from these two components using an effective three-step cyclization protocol. Thus, a highly convergent process for the drug candidate was defined.