Merck
Serum paraoxonase 1 activity is decreased in thyroid dysfunction.
Journal of endocrinological investigation (2003-12-13)
F Azizi, F Raiszadeh, M Solati, A Etemadi, M Rahmani, M Arabi
ABSTRACT
Changes in serum lipid and lipoprotein concentrations occur frequently in disorders of thyroid function. LDL-cholesterol (LDL-C) oxidation susceptibility is higher in these patients than in normal population. This study aims at assessing lipids, lipoproteins, apolipoproteins and serum paraoxonase 1 (PON1) activity in patients with thyroid dysfunction. Ninety-nine patients with thyroid dysfunction, (49 hypothyroid and 50 hyperthyroid) were compared with 2 separately age- and sex-matched control groups. A fasting blood sample was obtained and serum total cholesterol, triglycerides, apolipoproteins A-I and B, and PON1 activity were measured. In hyperthyroid patients, significantly lower PON1 activity (45 +/- 23 vs 67 +/- 37 IU/ml, p<0.001), triglycerides (112 +/- 53 vs 166 +/- 130 mg/dl, p<0.05), apolipoprotein A-I (137 +/- 26 vs 154 +/- 21 mg/dl, p<0.001) and apolipoprotein B (75 +/- 18 vs 86 +/- 25 mg/dl, p<0.05) were found. Hypothyroid patients had lower PON1 activity (46 +/- 21 vs 64 +/- 32 IU/ml, p<0.005) compared with controls, and higher total cholesterol (224 +/- 69 vs 185 +/- 41 mg/dl, p<0.001), LDL-C (133 +/- 59 vs 93 +/- 36 mg/dl, p<0.001), and apolipoprotein B (107 +/- 37 vs 84 +/- 23 mg/dl, p<0.001). The results show significant changes of lipid levels in thyroid dysfunction. In addition, a significant reduction in PON1 activity was observed in both hyper- and hypothyroid patients. Increased LDL-C oxidation in thyroid dysfunction observed in other studies, at least to some extent, can be attributed to reduced PON1 activity.