Merck
  • Active components from Artemisia iwayomogi displaying ONOO(-) scavenging activity.
Active components from Artemisia iwayomogi displaying ONOO(-) scavenging activity.
Phytotherapy research : PTR (2004-01-30)
Ae Ra Kim, Ya Ni Zou, Tae Hyun Park, Kyung Hee Shim, Min Sun Kim, Nam Deuk Kim, Jong Deog Kim, Song Ja Bae, Jae Sue Choi, Hae Young Chung
ABSTRACT
Peroxynitrite (ONOO(-)) is one of cytotoxic species produced by the reaction between superoxide (*O(2) (-)) and nitric oxide (NO). The main aim of this study was to identify ONOO(-) scavenging constituents from herbs. Methanolic extracts derived from one hundred thirty eight herbs were screened out for their ONOO(-) scavenging activities. Twenty herbs showed excellent ONOO(-) scavenging activity in a dose dependent manner. Of them, Artemisia iwayomogi, which showed the highest activity, was fractioned with several solvents. The ONOO(-) scavenging activity of fractions was in order of ethyl acetate (EtOAc) > n-butanol (BuOH) > dichloromethane (CH(2)Cl(2)) > water (H(2)O) fraction. The EtOAc and the n-BuOH fractions were further purified to isolate active principles by column chromatography. Apigenin 7-methylether (genkwanin), apigenin 7,4'-di-O methylether, jaceosidin, and scopoletin from the EtOAc fraction and chlorogenic acid, 2,4-dihydroxy 6-methoxy acetophenone 4-O-beta-D-glucoside, quebrachitol, and scopolin from the n-BuOH fraction were identified. The scavenging activities of chlorogenic acid (IC(50) = 0.52 +/- 0.04 microM), genkwanin (IC(50) = 1.01 +/- 0.10 microM), and scopoletin (IC(50) = 1.03 +/- 0.15 micro M) were higher than or comparable to a well-known ONOO(-) scavenger, penicillamine (IC(50) = 1.76 +/- 0.18 microM), suggesting that those compounds might be developed as effective ONOO(-) scavengers for, prevention of the ONOO(-)-involved diseases.
Sigma-Aldrich
Scopoletin, ≥99%