Merck
  • Podocalyxin is a glycoprotein ligand of the human pluripotent stem cell-specific probe rBC2LCN.
Podocalyxin is a glycoprotein ligand of the human pluripotent stem cell-specific probe rBC2LCN.
Stem cells translational medicine (2013-03-26)
Hiroaki Tateno, Asako Matsushima, Keiko Hiemori, Yasuko Onuma, Yuzuru Ito, Kayo Hasehira, Ken Nishimura, Manami Ohtaka, Satoko Takayasu, Mahito Nakanishi, Yuzuru Ikehara, Mio Nakanishi, Kiyoshi Ohnuma, Techuan Chan, Masashi Toyoda, Hidenori Akutsu, Akihiro Umezawa, Makoto Asashima, Jun Hirabayashi
ABSTRACT
In comprehensive glycome analysis with a high-density lectin microarray, we have previously shown that the recombinant N-terminal domain of the lectin BC2L-C from Burkholderia cenocepacia (rBC2LCN) binds exclusively to undifferentiated human induced pluripotent stem (iPS) cells and embryonic stem (ES) cells but not to differentiated somatic cells. Here we demonstrate that podocalyxin, a heavily glycosylated type 1 transmembrane protein, is a glycoprotein ligand of rBC2LCN on human iPS cells and ES cells. When analyzed by DNA microarray, podocalyxin was found to be highly expressed in both iPS cells and ES cells. Western and lectin blotting revealed that rBC2LCN binds to podocalyxin with a high molecular weight of more than 240 kDa in undifferentiated iPS cells of six different origins and four ES cell lines, but no binding was observed in either differentiated mouse feeder cells or somatic cells. The specific binding of rBC2LCN to podocalyxin prepared from a large set of iPS cells (138 types) and ES cells (15 types) was also confirmed using a high-throughput antibody-overlay lectin microarray. Alkaline digestion greatly reduced the binding of rBC2LCN to podocalyxin, indicating that the major glycan ligands of rBC2LCN are presented on O-glycans. Furthermore, rBC2LCN was found to exhibit significant affinity to a branched O-glycan comprising an H type 3 structure (Ka, 2.5 × 10(4) M(-1)) prepared from human 201B7 iPS cells, indicating that H type 3 is a most probable potential pluripotency marker. We conclude that podocalyxin is a glycoprotein ligand of rBC2LCN on human iPS cells and ES cells.
Product Description
Roche
Neuraminidase (Sialidase), from Arthrobacter ureafaciens
Pricing and availability is not currently available.