- Development of an air-stable nickel precatalyst for the amination of aryl chlorides, sulfamates, mesylates, and triflates.
Development of an air-stable nickel precatalyst for the amination of aryl chlorides, sulfamates, mesylates, and triflates.
Organic letters (2013-11-29)
Nathaniel H Park, Georgiy Teverovskiy, Stephen L Buchwald
PMID24283652
ABSTRACT
A new air-stable nickel precatalyst for C-N cross-coupling is reported. The developed catalyst system displays a greatly improved substrate scope for C-N bond formation to include both a wide range of aryl and heteroaryl electrophiles and aryl, heteroaryl, and alkylamines. The catalyst system is also compatible with a weak base, allowing the amination of substrates containing base-sensitive functional groups.