Merck
Importin beta: conducting a much larger cellular symphony.
Molecular cell (2004-11-05)
Amnon Harel, Douglass J Forbes
ABSTRACT
Importin beta, once thought to be exclusively a nuclear transport receptor, is emerging as a global regulator of diverse cellular functions. Importin beta acts positively in multiple interphase roles: in nuclear import, as a chaperone for highly charged nuclear proteins, and as a potential motor adaptor for movement along microtubules. In contrast, importin beta plays a negative regulatory role in mitotic spindle assembly, centrosome dynamics, nuclear membrane formation, and nuclear pore assembly. In most of these, importin beta is counteracted by its regulator, Ran-GTP. In light of this, the recent discovery of Ran's involvement in spindle checkpoint control suggested a potential new arena for importin beta action, although it is also possible that one of importin beta's relatives, the karyopherin family of proteins, manages this checkpoint. Lastly, importin beta plays a role in transducing damage signals from the axons of injured neurons back to the cell body.