- MicroRNA-224 sustains Wnt/β-catenin signaling and promotes aggressive phenotype of colorectal cancer.
MicroRNA-224 sustains Wnt/β-catenin signaling and promotes aggressive phenotype of colorectal cancer.
Growing evidence suggests that Wnt/β-catenin pathway plays an important role in CRC development, progression and metastasis. Aberrant miR-224 expression has been reported in CRC. However, the mechanism of miR-224 promotes both proliferation and metastatic ability largely remains unclear. Real-time PCR was used to quantify miR-224 expression. Luciferase reporter assays were conducted to confirm the activity of Wnt/β-catenin pathway and target gene associations, and immunofluorescence staining assay was performed to observe the nuclear translocation of β-catenin. Bioinformatics analysis combined with in vivo and vitro functional assays showed the potential target genes, GSK3β and SFRP2, of miR-224. Specimens from forty patients with CRC were analyzed for the expression of miR-224 and the relationship with GSK3β/SFRP2 by real-time PCR and western blot. Bioinformatics and cell luciferase function studies verified the direct regulation of miR-224 on the 3'-UTR of the GSK3β and SFRP2 genes, which leads to the activation of Wnt/β-catenin signaling and the nuclear translocation of β-catenin. In addition, knockdown of miR-224 significantly recovered the expression of GSK3β and SFRP2 and attenuated Wnt/β-catenin-mediated cell metastasis and proliferation. The ectopic upregulation of miR-224 dramatically inhibited the expression of GSK3β/SFRP2 and enhanced CRC proliferation and invasion. Our research showed mechanistic links between miR-224 and Wnt/β-catenin in the pathogenesis of CRC through modulation of GSK3β and SFRP2.