Merck
  • Structure, function, property, and role in neurologic diseases and other diseases of the sHsp22.
Structure, function, property, and role in neurologic diseases and other diseases of the sHsp22.
Journal of neuroscience research (2007-02-17)
Zhiping Hu, Lan Chen, Jie Zhang, Ting Li, Jianguang Tang, Niangui Xu, Xiang Wang
ABSTRACT
Small heat shock proteins are members of the heat shock proteins family. They share important identical features: 1) they form the conserved structure 'alpha-crystallin domain' with about 80-100 residues in the C-terminal part of the proteins; 2) they have monomeric molecular masses ranging in 12-43 kDa; 3) they associate into large oligomers consisting in many cases of subunits; 4) they increase expression under stress conditions; 5) they exhibit a highly dynamic structure; and 6) they play a chaperone-like role. Hsp22 (also known as HspB8, H11, and E2IG1) retains the structural motif of the 'alpha-crystallin' family of Hsps and is a member of the superfamily of sHsps. Hsp22 displays chaperone activity, autokinase activity, and trigger or block apoptosis activity. It differs from canonical family members existing as a monomer. A decrease in the HspB8 activity may contribute to the development of some neurologic diseases and others.