Merck
  • Effect of para-sulfonato-calix[n]arenes on the solubility, chemical stability, and bioavailability of a water insoluble drug nifedipine.
Effect of para-sulfonato-calix[n]arenes on the solubility, chemical stability, and bioavailability of a water insoluble drug nifedipine.
Current drug discovery technologies (2008-08-05)
Wenzhan Yang, Daniel P Otto, Wilna Liebenberg, Melgardt M de Villiers
ABSTRACT
This study reports the use of para-sulphonato calix[8]arene to produce stable complexes with improved bioavailability for nifedipine, a calcium-channel blocker that is practically insoluble in water. Thermal analysis and electrospray ionisation mass spectroscopy confirmed that nifedipine formed complexes with the calixarenes in a size dependent way. The most stable, soluble complexes was formed with para-sulphonato calix[8]arene. Complexation was weakest with the calix[4]arene while complexation with the calix[6]arene was intermediate. However, the calix[4 and 6]arenes changed the chemical stability of the drug in solution because significant amounts of the nitroso-pyridine derivative was produced, proposing an interaction between the nifedipine bearing a H substituent at the N-1 position and the calixarenes. This oxidative degradation of the drug was greatest when combined with the calix[6]arene. Simultaneous oral ingestion of the calix[6 or 8]arenes significantly increased the bioavailability of the drug after oral administration in male Sprague-Dawley rats while not influencing CYP3A activities in the liver. The pharmacokinetic parameters of the nifedipine: para-sulfonato calix[8]arene complexes showed it was bioequivalent to a nifedipine PEG-solution. The absolute bioavailability for both formulations was ca. 60 %.
Sigma-Aldrich
Oxidized Nifedipine, powder, ~95% (HPLC)
USP
Nifedipine Nitrophenylpyridine Analog, United States Pharmacopeia (USP) Reference Standard