Merck
  • The association of mid-regional pro-adrenomedullin and mid-regional pro-atrial natriuretic peptide with mortality in an incident dialysis cohort.
The association of mid-regional pro-adrenomedullin and mid-regional pro-atrial natriuretic peptide with mortality in an incident dialysis cohort.
PloS one (2011-03-17)
Ghazaleh Gouya, Gisela Sturm, Claudia Lamina, Emanuel Zitt, Otto Freistätter, Joachim Struck, Michael Wolzt, Florian Knoll, Friederike Lins, Karl Lhotta, Ulrich Neyer, Florian Kronenberg
ABSTRACT
High levels of the plasma peptides mid-regional pro-adrenomedullin (MR-proADM) and mid-regional pro-atrial natriuretic peptide (MR-proANP) are associated with clinical outcomes in the general population. Data in patients with chronic kidney disease are sparse. We therefore investigated the association of MR-proANP and MR-proADM levels with all-cause and cardiovascular (CV) mortality, CV events and peripheral arterial disease in 201 incident dialysis patients of the INVOR-Study prospectively followed for a period of up to more than 7 years. The overall mortality rate was 43%, thereof 43% due to CV events. Both baseline MR-proANP and MR-proADM were associated with higher risk of all-cause (HR = 1.44, p = 0.001 and HR = 1.32, p = 0.002, respectively) and CV mortality (HR = 1.75, p<0.001 and HR = 1.41, p = 0.007, respectively) after adjustment for age, sex, previous CV events, diabetes mellitus and time-dependent type of renal replacement therapy. We then stratified patients in high risk (both peptides in the upper tertile), intermediate risk (only one of the two peptides in the upper tertile) and low risk (none in the upper tertile). Although demographic, clinical and laboratory variables were similar among the intermediate and high risk group, to be with both parameters in the upper tertile was associated with a 3-fold higher risk for all-cause (HR = 2.87, p<0.001) and CV mortality (HR = 3.58, p = 0.001). In summary, among incident dialysis patients MR-proANP and MR-proADM were shown to be associated with all-cause and CV mortality, with the highest risk when both parameters were in the upper tertiles.